Learn More
Strem, An Ascensus Company CAS# 67579-81-1. 2g. trans-N,N'-Dimethyl-1,2-cyclohexanediamine, 98%. MFCD03001702
SDP
Catalog No. 5090108914
Encompass
CAS# 67579-81-1. 2g. trans-N,N'-Dimethyl-1,2-cyclohexanediamine, 98%. MFCD03001702. Molecular Weight: 142.24. Molecular Formula: C6H10(NHCH3)2. Color/form: colorless to pale yellow liq. Strem# 07-0270. http://www.strem.com/catalog/v/07-0270/
Catalog No. 50-901-08914 Supplier Strem, An Ascensus Company Supplier No. 0702702G
CAS# 67579-81-1. 2g. trans-N,N'-Dimethyl-1,2-cyclohexanediamine, 98%. MFCD03001702. Molecular Weight: 142.24. Molecular Formula: C6H10(NHCH3)2. Color/form: colorless to pale yellow liq. Strem# 07-0270. http://www.strem.com/catalog/v/07-0270/