Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline 98.0+%, TCI America™
SDP

Catalog No. B16355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16355G Supplier TCI America Supplier No. B16355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13726-69-7
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD00053370
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 88804
IUPAC Name 1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.