Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-Ned 19 | 1354235-96-3 | MFCD02057367 | 99.3% | 514.6 g/mol | C30H31FN4O3 | 1 ML
SDP

Catalog No. 5000247755
Encompass_Preferred

trans-Ned 19 is a nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist that blocks two-pore channels and suppresses intracellular calcium signaling. It is supplied as a concentrated solution intended for research use only.

  • Supplied as a 10 mM solution in DMSO
  • Packaged volume: 1 mL
  • High purity, approximately 99.3%
  • Molecular weight: 514.59 g/mol
  • Chemical formula: C30H31FN4O3
  • Used to modulate calcium-dependent cellular processes in research

Catalog No. 50-002-47755 Supplier Medchemexpress LLC Supplier No. HY10331610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Ned 19 is a nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist that blocks two-pore channels and suppresses intracellular calcium signaling. It is supplied as a concentrated solution intended for research use only.

  • Supplied as a 10 mM solution in DMSO
  • Packaged volume: 1 mL
  • High purity, approximately 99.3%
  • Molecular weight: 514.59 g/mol
  • Chemical formula: C30H31FN4O3
  • Used to modulate calcium-dependent cellular processes in research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.