Learn More
Medchemexpress LLC Trans-Ned 19 | 1354235-96-3 | MFCD20926336 | 99.3% | 514.59 g/mol | C30H31FN4O3 | 50 MG
SDP
Catalog No. 5000244685
Encompass_Preferred
trans-Ned 19 is a nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist and two-pore channel (TPC) blocker used in calcium signaling research. It inhibits NAADP-mediated Ca2+ release (reported IC50 ≈ 6 nM) and reduces calcium signals in cellular assays. Chemical identifiers: CAS 1354235-96-3; formula C30H31FN4O3; molecular weight 514.59 g/mol.
  • Acts as an NAADP antagonist and two-pore channel blocker.
  • Inhibits NAADP-mediated Ca2+ release (IC50 ≈ 6 nM).
  • High purity for biochemical and cellular assays.
  • Supplied as a 50 mg solid; store at -20°C.
Catalog No. 50-002-44685 Supplier Medchemexpress LLC Supplier No. HY10331650MG
trans-Ned 19 is a nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist and two-pore channel (TPC) blocker used in calcium signaling research. It inhibits NAADP-mediated Ca2+ release (reported IC50 ≈ 6 nM) and reduces calcium signals in cellular assays. Chemical identifiers: CAS 1354235-96-3; formula C30H31FN4O3; molecular weight 514.59 g/mol.
  • Acts as an NAADP antagonist and two-pore channel blocker.
  • Inhibits NAADP-mediated Ca2+ release (IC50 ≈ 6 nM).
  • High purity for biochemical and cellular assays.
  • Supplied as a 50 mg solid; store at -20°C.