Learn More
Medchemexpress LLC Trans-Ned 19 | 1354235-96-3 | MFCD31562774 | 99.3% | 514.59 | C30H31FN4O3 | 5 MG
SDP
Catalog No. 5000247739
Encompass_Preferred
trans-Ned 19 is a small-molecule NAADP antagonist and two-pore channel (TPC) blocker used in preclinical research to modulate intracellular calcium signaling. Supplied as a high-purity solid and as a DMSO solution, it is intended for biochemical and cellular assays.
Catalog No. 50-002-47739 Supplier Medchemexpress LLC Supplier No. HY1033165MG
trans-Ned 19 is a small-molecule NAADP antagonist and two-pore channel (TPC) blocker used in preclinical research to modulate intracellular calcium signaling. Supplied as a high-purity solid and as a DMSO solution, it is intended for biochemical and cellular assays.