Learn More
Medchemexpress LLC Trans-Ned 19 | 1354235-96-3 | MFCD31562774 | 99.3% | 514.59 g/mol | C30H31FN4O3 | 10 MG
SDP
Catalog No. 5000244827
Encompass_Preferred
trans-Ned 19 is a research chemical that functions as a nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist and two-pore channel (TPC) blocker, inhibiting NAADP-mediated calcium (Ca2+) release. It is supplied as a solid for laboratory research use and is characterized by high purity suitable for analytical and biological assays.
  • Acts as an NAADP antagonist and TPC blocker.
  • Inhibits NAADP-mediated Ca2+ release (reported IC50 ≈ 6 nM).
  • High purity appropriate for analytical and biological experiments.
  • Supplied as a solid, stable under recommended storage conditions.
  • Molecular weight 514.59 g/mol; chemical formula C30H31FN4O3.
  • Available in small-milligram quantities for research applications.
Catalog No. 50-002-44827 Supplier Medchemexpress LLC Supplier No. HY10331610MG
trans-Ned 19 is a research chemical that functions as a nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist and two-pore channel (TPC) blocker, inhibiting NAADP-mediated calcium (Ca2+) release. It is supplied as a solid for laboratory research use and is characterized by high purity suitable for analytical and biological assays.
  • Acts as an NAADP antagonist and TPC blocker.
  • Inhibits NAADP-mediated Ca2+ release (reported IC50 ≈ 6 nM).
  • High purity appropriate for analytical and biological experiments.
  • Supplied as a solid, stable under recommended storage conditions.
  • Molecular weight 514.59 g/mol; chemical formula C30H31FN4O3.
  • Available in small-milligram quantities for research applications.