Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-permethrin | 61949-77-7 | MFCD00145431 | 99.9% | 391.29 g·mol⁻¹ | C21H20Cl2O3 | 25 MG
SDP

Catalog No. 5000284023
Encompass_Preferred

trans-Permethrin is the trans stereoisomer of permethrin, a synthetic pyrethroid insecticide supplied for research use. It is used in studies of insecticidal activity and neurotoxicity, and is provided in milligram-scale packages suitable for laboratory experiments.

  • High purity (99.88%).
  • Molecular formula C21H20Cl2O3.
  • Molecular weight 391.29 g·mol⁻¹.
  • Available in mg-scale pack sizes (for example: 5 mg, 10 mg, 25 mg).
  • Intended for research use only; not for human or veterinary use.
  • Suitable for biological, toxicological, and insecticide efficacy studies.

Catalog No. 50-002-84023 Supplier Medchemexpress LLC Supplier No. HYW58821225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Permethrin is the trans stereoisomer of permethrin, a synthetic pyrethroid insecticide supplied for research use. It is used in studies of insecticidal activity and neurotoxicity, and is provided in milligram-scale packages suitable for laboratory experiments.

  • High purity (99.88%).
  • Molecular formula C21H20Cl2O3.
  • Molecular weight 391.29 g·mol⁻¹.
  • Available in mg-scale pack sizes (for example: 5 mg, 10 mg, 25 mg).
  • Intended for research use only; not for human or veterinary use.
  • Suitable for biological, toxicological, and insecticide efficacy studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.