Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-permethrin | 61949-77-7 | MFCD00145431 | 99.9% | 391.29 g·mol⁻¹ | C21H20Cl2O3 | 5 MG
SDP

Catalog No. 5000284005
Encompass_Preferred

Trans-permethrin is the trans isomer of permethrin, a synthetic pyrethroid insecticide used in research as a neurotoxin and mitochondrial complex I inhibitor. The material is supplied as a solid for laboratory use and is intended for research only.

  • High purity (99.9%).
  • White to off-white solid appearance.
  • Available in small quantities suitable for analytical and biochemical assays.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder at -20°C for long term, 4°C for shorter term.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-84005 Supplier Medchemexpress LLC Supplier No. HYW5882125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trans-permethrin is the trans isomer of permethrin, a synthetic pyrethroid insecticide used in research as a neurotoxin and mitochondrial complex I inhibitor. The material is supplied as a solid for laboratory use and is intended for research only.

  • High purity (99.9%).
  • White to off-white solid appearance.
  • Available in small quantities suitable for analytical and biochemical assays.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder at -20°C for long term, 4°C for shorter term.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.