Learn More
Thomas Scientific Transfer Pipet, 3.0mL, Small Bulb, Graduated to 1mL, 140mm, STERILE, Individually Wrapped, Cellophane Wrap, Bulb Draw - 2.2mL
SDP
Catalog No. NC1868898
Encompass
Transfer Pipet, 3.0mL, Small Bulb, Graduated to 1mL, 140mm, STERILE, Individually Wrapped, Cellophane Wrap, Bulb Draw - 2.2mL. Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean.
Catalog No. NC1868898 Supplier Thomas Scientific Supplier No. 1193V27
Transfer Pipet, 3.0mL, Small Bulb, Graduated to 1mL, 140mm, STERILE, Individually Wrapped, Cellophane Wrap, Bulb Draw - 2.2mL. Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean.