Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarstedt Inc Transfer Pipette, 3.5mL, Narrow, 500/Pack

Catalog No. NC9901373 Shop All Sarstedt Inc. Products
Encompass

Transfer Pipette 3.5ml Narrow (Encompass)

Catalog No. NC9901373 Supplier Sarstedt Inc Supplier No. 86.1173.300
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Transfer Pipette 3.5ml Narrow (Encompass)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.