Learn More
Medchemexpress LLC trans-Pralsetinib | 2097132-93-7 | 533.60 | 1 ML
SDP
Catalog No. 5000438461
Encompass_Preferred
trans-Pralsetinib (trans-BLU-667) is a rearranged during transfection (RET) inhibitor, derived from patent US20170121312A1, Compound Example 129.
  • Potent rearranged during transfection (RET) inhibitor
  • Targets the RET pathway
  • Relevant for protein tyrosine kinase/RTK research
Catalog No. 50-004-38461 Supplier Medchemexpress LLC Supplier No. HY112301A10MM/1ML
trans-Pralsetinib (trans-BLU-667) is a rearranged during transfection (RET) inhibitor, derived from patent US20170121312A1, Compound Example 129.
  • Potent rearranged during transfection (RET) inhibitor
  • Targets the RET pathway
  • Relevant for protein tyrosine kinase/RTK research