Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-PX20606 (trans-PX-102) | 1268244-85-4 | 99.2% | 554.85 | C29H22Cl3NO4 | 1 ML
SDP

Catalog No. 5000214930
Encompass_Preferred

trans-PX20606 (trans-PX-102) is a small-molecule agonist of the farnesoid X receptor (FXR) for research use. It exhibits low-nanomolar potency in FXR functional assays and is supplied as a 10 mM solution in DMSO or as solid quantities for biochemical and cellular studies.

  • Low-nanomolar FXR agonist activity with reported EC50s around 32-34 nM.
  • Available as 10 mM solution in DMSO and as solid quantities (2 mg, 5 mg, 10 mg).
  • High chemical purity, typically ≥99.2% by HPLC.
  • Molecular weight approximately 554.85 g·mol⁻¹ and formula C29H22Cl3NO4.
  • Intended for in vitro research applications; not for human or veterinary use.

Catalog No. 50-002-14930 Supplier Medchemexpress LLC Supplier No. HY100443A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-PX20606 (trans-PX-102) is a small-molecule agonist of the farnesoid X receptor (FXR) for research use. It exhibits low-nanomolar potency in FXR functional assays and is supplied as a 10 mM solution in DMSO or as solid quantities for biochemical and cellular studies.

  • Low-nanomolar FXR agonist activity with reported EC50s around 32-34 nM.
  • Available as 10 mM solution in DMSO and as solid quantities (2 mg, 5 mg, 10 mg).
  • High chemical purity, typically ≥99.2% by HPLC.
  • Molecular weight approximately 554.85 g·mol⁻¹ and formula C29H22Cl3NO4.
  • Intended for in vitro research applications; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.