Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Quercus lactone, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014133 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4133 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4133 Supplier MilliporeSigma Supelco Supplier No. 72777100MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 39638-67-0
Linear Formula C9H16O2
MDL Number MFCD28010977
Synonym trans-5-Butyl-4-methyloxolan-2-one; trans-5-Butyldihydro-4-methyl-2(3 H)-furanone; trans-Methyloctanolide; trans-Oak lactone; trans-Whiskey lactone; trans-beta-Methyl-gamma-octalactone
Type Analytical Standard
Quantity 100 mg
Formula Weight 156.22
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.