Learn More
Medchemexpress LLC Trans-R-138727 | 239466-74-1 | 99.0% | 349.42 g/mol | C18H20FNO3S | 1MG
SDP
Catalog No. 5000237559
Encompass_Preferred
Trans-R-138727 is the trans isomer of R-138727, the thiol-containing active metabolite of the antiplatelet agent prasugrel. It is supplied as a high-purity research chemical for biochemical and pharmacological studies, including platelet P2Y12 receptor inhibition assays and metabolite characterization.
  • High purity suitable for analytical and biological assays.
  • Useful as a reference standard and pharmacological tool.
  • Provided as a powder with low-temperature storage recommendations.
  • Molecular weight 349.42 g/mol and formula C18H20FNO3S.
  • Available in milligram-scale quantities for research use.
Catalog No. 50-002-37559 Supplier Medchemexpress LLC Supplier No. HY123669A1MG
Trans-R-138727 is the trans isomer of R-138727, the thiol-containing active metabolite of the antiplatelet agent prasugrel. It is supplied as a high-purity research chemical for biochemical and pharmacological studies, including platelet P2Y12 receptor inhibition assays and metabolite characterization.
  • High purity suitable for analytical and biological assays.
  • Useful as a reference standard and pharmacological tool.
  • Provided as a powder with low-temperature storage recommendations.
  • Molecular weight 349.42 g/mol and formula C18H20FNO3S.
  • Available in milligram-scale quantities for research use.