Learn More
Medchemexpress LLC Trans-R-138727 | 239466-74-1 | 99.0% | 349.42 g/mol | C18H20FNO3S | 25MG
SDP
Catalog No. 5000236714
Encompass_Preferred
trans-R-138727 is the trans stereoisomer of the active metabolite of prasugrel. It is a thiol-containing compound that irreversibly inhibits the platelet P2Y12 receptor and is intended for use as a research reference in studies of platelet aggregation and antiplatelet pharmacology. Handle according to applicable laboratory safety and waste procedures.
  • Irreversible P2Y12 receptor inhibitor.
  • Thiol-containing active metabolite of prasugrel.
  • High-purity solid suitable for biochemical assays.
  • Molecular formula C18H20FNO3S; molecular weight 349.42 g/mol.
  • Available in small pack sizes for research use.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-36714 Supplier Medchemexpress LLC Supplier No. HY123669A25MG
trans-R-138727 is the trans stereoisomer of the active metabolite of prasugrel. It is a thiol-containing compound that irreversibly inhibits the platelet P2Y12 receptor and is intended for use as a research reference in studies of platelet aggregation and antiplatelet pharmacology. Handle according to applicable laboratory safety and waste procedures.
  • Irreversible P2Y12 receptor inhibitor.
  • Thiol-containing active metabolite of prasugrel.
  • High-purity solid suitable for biochemical assays.
  • Molecular formula C18H20FNO3S; molecular weight 349.42 g/mol.
  • Available in small pack sizes for research use.
  • For research use only; not for human therapeutic use.