Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Sinapic acid, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015383 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5383 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5383 Supplier MilliporeSigma Supelco Supplier No. 9387850MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 7362-37-0
Physical Form Neat
Linear Formula C11H12O5
MDL Number MFCD00004401
Melting Point 195°C to 205°C
Synonym trans-4-Hydroxy-3,5-dimethoxy-cinnamic acid; trans-Sinapinic acid
Type Analytical Standard
Quantity 50 mg
Formula Weight 224.21
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.