Learn More
Cayman Chemical trans-ISRIB, Purity: ≥95%, CAS: 1597403-47-8, 5 mg
SDP
Catalog No. 502971331
Encompass_Preferred
A symmetric bis-glycolamide that inhibits the eIF2α phosphorylation-mediated unfolded protein response (IC50 = 5 nM in a cell-reporter assay); passes through the blood-brain barrier and significantly enhances learning and memory in mice and rats
Catalog No. 50-297-1331 Supplier Cayman Chemical Supplier No. 162585 MG
A symmetric bis-glycolamide that inhibits the eIF2α phosphorylation-mediated unfolded protein response (IC50 = 5 nM in a cell-reporter assay); passes through the blood-brain barrier and significantly enhances learning and memory in mice and rats