Learn More
Medchemexpress LLC trans-Stilbene | 103-30-0 | 100.0% | 1 G
SDP
Catalog No. 5000342328
Encompass_Preferred
trans-Stilbene ((E)-Stilbene) is used in the manufacturing of dye lasers, optical brighteners, and non-steroidal synthetic estrogens. It exhibits weak antioxidant activity against APPH-induced free radicals and also shows weak reducing effects against ADP-Fe3+ and EDTA-Fe3+.
  • Utilized in dye laser production.
  • Used for optical brighteners.
  • A component for non-steroidal synthetic estrogens.
  • Demonstrates weak antioxidant activity.
  • Shows weak reducing effects.
Catalog No. 50-003-42328 Supplier Medchemexpress LLC Supplier No. HY1287931G
trans-Stilbene ((E)-Stilbene) is used in the manufacturing of dye lasers, optical brighteners, and non-steroidal synthetic estrogens. It exhibits weak antioxidant activity against APPH-induced free radicals and also shows weak reducing effects against ADP-Fe3+ and EDTA-Fe3+.
  • Utilized in dye laser production.
  • Used for optical brighteners.
  • A component for non-steroidal synthetic estrogens.
  • Demonstrates weak antioxidant activity.
  • Shows weak reducing effects.