Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Stilbene, 96%, Thermo Scientific Chemicals

Catalog No. ac161041000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161041000 100 g Plastic bottle
AC161045000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161041000 Supplier Thermo Scientific Chemicals Supplier No. 161041000
Only null left
Add to Cart

trans-Stilbene, 96%, C14H12, CAS Number-103-30-0, bibenzylidine, trans-stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, alpha,beta-diphenylethylene, stilbene, trans-1,2-diphenylethene, bibenzal, e-stilbene, bibenzylidene, 500g, 95% min. (HPLC), 05,II,537

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-30-0
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00064300
InChI Key PJANXHGTPQOBST-VAWYXSNFSA-N
Synonym trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
PubChem CID 638088
ChEBI CHEBI:36007
IUPAC Name (E)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 122°C to 126°C
Color White to Beige
Boiling Point 305°C to 307°C (744.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CH=CHC6H5
Quantity 100 g
Beilstein 05,II,537
Merck Index 15,8946
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 90 parts cold alcohol,freely soluble in benzene and ether
Formula Weight 180.25
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-Stilbene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-098-5
RTECSNumber WJ4926500
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.