Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-Sulfo-SMCC | 1286837-77-1 | 90.9% | 414.39 | C16H18N2O9S | 10 MG
SDP

Catalog No. 5000203740
Encompass_Preferred

trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker commonly used as an ADC linker reagent in conjugation workflows.

  • Non-cleavable, membrane-permeable crosslinker suitable for ADC applications.
  • CAS number 1286837-77-1 for unambiguous identification.
  • Molecular formula C16H18N2O9S and molecular weight 414.39 g/mol.
  • Typical purity around 90.9% as supplied.
  • Available in small-milligram quantities for research use.

Catalog No. 50-002-03740 Supplier Medchemexpress LLC Supplier No. HY12650310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker commonly used as an ADC linker reagent in conjugation workflows.

  • Non-cleavable, membrane-permeable crosslinker suitable for ADC applications.
  • CAS number 1286837-77-1 for unambiguous identification.
  • Molecular formula C16H18N2O9S and molecular weight 414.39 g/mol.
  • Typical purity around 90.9% as supplied.
  • Available in small-milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.