Learn More
Medchemexpress LLC trans-Sulfo-SMCC | 1286837-77-1 | 90.9% | 414.39 g·mol⁻1 | C16H18N2O9S | 5 MG
SDP
Catalog No. 5000202458
Encompass_Preferred
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker containing an NHS ester and a maleimide reactive group. It is supplied as a solid for research use in bioconjugation and ADC development.
  • Non-cleavable crosslinker for stable conjugation
  • Membrane-permeable design suitable for intracellular labeling
  • Reacts with primary amines and sulfhydryls via NHS ester and maleimide
  • Supplied as an off-white to white solid powder
  • Stable when stored at -20°C in powder form
Catalog No. 50-002-02458 Supplier Medchemexpress LLC Supplier No. HY1265035MG
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker containing an NHS ester and a maleimide reactive group. It is supplied as a solid for research use in bioconjugation and ADC development.
  • Non-cleavable crosslinker for stable conjugation
  • Membrane-permeable design suitable for intracellular labeling
  • Reacts with primary amines and sulfhydryls via NHS ester and maleimide
  • Supplied as an off-white to white solid powder
  • Stable when stored at -20°C in powder form