Learn More
Medchemexpress LLC Trans-sulfo-SMCC | 1286837-77-1 | 90.9% | 414.39 g/mol | C16H18N2O9S | 100 MG
SDP
Catalog No. 5000204045
Encompass_Preferred
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable crosslinker used for antibody-drug conjugate (ADC) construction and general bioconjugation. It contains maleimide and N-hydroxysuccinimide (NHS) ester functionalities that enable stable conjugation between sulfhydryl and amino groups to form non-cleavable linkages.
  • Non-cleavable linker suitable for ADC construction.
  • Membrane permeable for potential intracellular delivery.
  • Maleimide and NHS-ester reactive groups for thiol-to-amine conjugation.
  • Solid, white to off-white, available in multiple quantities including 100 mg.
  • Stable as a powder at -20 °C for long-term storage; in solvent stability varies.
Catalog No. 50-002-04045 Supplier Medchemexpress LLC Supplier No. HY126503100MG
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable crosslinker used for antibody-drug conjugate (ADC) construction and general bioconjugation. It contains maleimide and N-hydroxysuccinimide (NHS) ester functionalities that enable stable conjugation between sulfhydryl and amino groups to form non-cleavable linkages.
  • Non-cleavable linker suitable for ADC construction.
  • Membrane permeable for potential intracellular delivery.
  • Maleimide and NHS-ester reactive groups for thiol-to-amine conjugation.
  • Solid, white to off-white, available in multiple quantities including 100 mg.
  • Stable as a powder at -20 °C for long-term storage; in solvent stability varies.