Learn More
Medchemexpress LLC trans-Sulfo-SMCC | 1286837-77-1 | 90.9% | 414.39 g/mol | C16H18N2O9S | 25 MG
SDP
Catalog No. 5000202199
Encompass_Preferred
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker containing maleimide and N-hydroxysuccinimide (NHS) ester reactive groups for conjugation to sulfhydryl and amine groups. It is supplied as a solid for research use in bioconjugation and ADC linker development.
  • Non-cleavable linker suitable for stable conjugation
  • Membrane permeable to support intracellular delivery
  • Maleimide and NHS-ester reactive groups for sulfhydryl and amine coupling
  • Available in multiple pack sizes, including 25 mg
  • Supplied with purity approximately 90.9%
  • Molecular weight ~414.39 g/mol
Catalog No. 50-002-02199 Supplier Medchemexpress LLC Supplier No. HY12650325MG
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker containing maleimide and N-hydroxysuccinimide (NHS) ester reactive groups for conjugation to sulfhydryl and amine groups. It is supplied as a solid for research use in bioconjugation and ADC linker development.
  • Non-cleavable linker suitable for stable conjugation
  • Membrane permeable to support intracellular delivery
  • Maleimide and NHS-ester reactive groups for sulfhydryl and amine coupling
  • Available in multiple pack sizes, including 25 mg
  • Supplied with purity approximately 90.9%
  • Molecular weight ~414.39 g/mol