Learn More
Medchemexpress LLC Trans-sulfo-SMCC | 1286837-77-1 | 90.9% | 414.39 g/mol | C16H18N2O9S | 50 MG
SDP
Catalog No. 5000202208
Encompass_Preferred
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker used to form stable, non-cleavable linkages between proteins and small-molecule payloads in research and bioconjugation workflows. It contains functional groups reactive toward primary amines and sulfhydryls, enabling a range of conjugation strategies.
  • Non-cleavable linker for producing stable conjugates.
  • Membrane permeable to facilitate intracellular delivery of conjugated payloads.
  • Reactive toward primary amines and sulfhydryls for flexible conjugation chemistry.
  • Available in multiple sizes; commonly supplied as 50 MG vials.
  • Purity approximately 90.9% as reported on the product page.
  • Solid, white to off-white appearance; store powder at -20°C for long-term stability.
Catalog No. 50-002-02208 Supplier Medchemexpress LLC Supplier No. HY12650350MG
trans-Sulfo-SMCC is a non-cleavable, membrane-permeable antibody-drug conjugate (ADC) crosslinker used to form stable, non-cleavable linkages between proteins and small-molecule payloads in research and bioconjugation workflows. It contains functional groups reactive toward primary amines and sulfhydryls, enabling a range of conjugation strategies.
  • Non-cleavable linker for producing stable conjugates.
  • Membrane permeable to facilitate intracellular delivery of conjugated payloads.
  • Reactive toward primary amines and sulfhydryls for flexible conjugation chemistry.
  • Available in multiple sizes; commonly supplied as 50 MG vials.
  • Purity approximately 90.9% as reported on the product page.
  • Solid, white to off-white appearance; store powder at -20°C for long-term stability.