Learn More
TARGETMOL CHEMICALS INC TRANS-TRANILAST 25MG
SDP
Catalog No. 502580934
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trans-Tranilast is an antiallergic drug developed by Kissei Pharmaceuticals. It was approved in 1982 for use in Japan and South Korea for bronchial asthma. Indications for keloid and hypertrophic scar were added in 1993. It has been used for the treatment of allergic disorders such as asthma allergic rhinitis and atopic dermatitis. purity: 99%
Catalog No. 50-258-0934 Supplier TARGETMOL CHEMICALS INC Supplier No. T227025MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trans-Tranilast is an antiallergic drug developed by Kissei Pharmaceuticals. It was approved in 1982 for use in Japan and South Korea for bronchial asthma. Indications for keloid and hypertrophic scar were added in 1993. It has been used for the treatment of allergic disorders such as asthma allergic rhinitis and atopic dermatitis. purity: 99%