Learn More
Medchemexpress LLC trans-Tranilast | 70806-55-2 | 99.0% | 327.34 | 5 MG
SDP
Catalog No. 5000387759
Encompass_Preferred
trans-Tranilast (trans-MK-341) is an anti-allergy agent that is utilized in research studies focusing on bronchial asthma, allergic rhinitis, and atopic dermatitis.
  • Functions as an anti-allergy agent.
  • Applicable in studies related to bronchial asthma.
  • Used in research concerning allergic rhinitis.
  • Investigated for its role in atopic dermatitis studies.
  • High purity of 98.99%.
  • Available in solid form.
Catalog No. 50-003-87759 Supplier Medchemexpress LLC Supplier No. HY187065MG
trans-Tranilast (trans-MK-341) is an anti-allergy agent that is utilized in research studies focusing on bronchial asthma, allergic rhinitis, and atopic dermatitis.
  • Functions as an anti-allergy agent.
  • Applicable in studies related to bronchial asthma.
  • Used in research concerning allergic rhinitis.
  • Investigated for its role in atopic dermatitis studies.
  • High purity of 98.99%.
  • Available in solid form.