Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one 98.0+%, TCI America™
SDP

Catalog No. B22835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22835G Supplier TCI America Supplier No. B22835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53369-00-9
Molecular Formula C17H12F2O
Molecular Weight (g/mol) 270.279
MDL Number MFCD00298485
InChI Key BNHFGYIPXPENKA-YDWXAUTNSA-N
Synonym trans,trans-Bis(4-fluorobenzal)acetone, trans,trans-Bis(4-fluorobenzylidene)acetone, trans,trans-Bis(4-fluorostyryl) Ketone
PubChem CID 1268251
IUPAC Name (1E,4E)-1,5-bis(4-fluorophenyl)penta-1,4-dien-3-one
SMILES C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)F)F

Specifications

Melting Point 154°C
Quantity 5 g
Formula Weight 270.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.