Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-2,4-Decadien-1-ol, 90%, remainder mainly trans, cis isomer

Catalog No. AAA1598903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1598903 1 g
AAA1598906 5 g
AAA1598914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1598903 Supplier Thermo Scientific Chemicals Supplier No. A1598903
Add to Cart

CAS: 18409-21-7 | C10H18O | 154.25 g/mol

trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.

Solubility
Not miscible in water. Soluble in fat.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18409-21-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00014052
InChI Key NUBWFSDCZULDCI-BLHCBFLLSA-N
Synonym 2,4-decadien-1-ol, 2e,4e-2,4-decadien-1-ol, 2e,4e-deca-2,4-dien-1-ol, e,e-2,4-decadien-1-ol, unii-21kkw523r4, trans,trans-2,4-decadien-1-ol, trans,trans-2,4-decadienol, 2,4-decadien-1-ol, 2e,4e, trans-2,4-decadienol, 2,4-decadien-1-ol, e,e
PubChem CID 5362696
IUPAC Name (2E,4E)-deca-2,4-dien-1-ol
SMILES CCCCC\C=C\C=C\CO

Specifications

Density 0.87
Boiling Point 80°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.489
Quantity 1 g
Solubility Information Not miscible in water. Soluble in fat.
Formula Weight 154.26
Percent Purity 90%
Assay remainder mainly trans, cis isomer
Chemical Name or Material trans,trans-2,4-Decadien-1-ol
EINECSNumber 242-291-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.