Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans,trans-2,4-Decadienal | 25152-84-5 | 99.9% | 152.23 | 1 G
SDP

Catalog No. 5000454465
Encompass_Preferred

trans,trans-2,4-Decadienal is a lipid peroxidation product of linoleic acid. It is for research use only and available as a liquid.

  • Purity: 99.9%
  • Appearance: Liquid
  • Color: Light yellow to green yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (pure form): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-54465 Supplier Medchemexpress LLC Supplier No. HYW0136271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans,trans-2,4-Decadienal is a lipid peroxidation product of linoleic acid. It is for research use only and available as a liquid.

  • Purity: 99.9%
  • Appearance: Liquid
  • Color: Light yellow to green yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (pure form): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.