Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans,trans-2,4-Decadienal | 25152-84-5 | 99.9% | 152.23 | 1 ML
SDP

Catalog No. 5000454658
Encompass_Preferred

Trans,trans-2,4-Decadienal, also known by its synonym (2E,4E)-癸-2,4-二烯醛, is a lipid peroxidation product derived from linoleic acid. This compound has a molecular weight of 152.23 and the chemical formula C10H16O. It is a liquid with a density of 0.854±0.06 g/cm3, appearing light yellow to green yellow. Primarily used for scientific research or pharmaceutical registration.

  • Derived from linoleic acid
  • Molecular weight of 152.23
  • Light yellow to green yellow liquid
  • Used for scientific research or pharmaceutical registration
  • Metabolized via oxidation or glutathione conjugation
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at -20°C for pure form; -80°C or -20°C in solvent

Catalog No. 50-004-54658 Supplier Medchemexpress LLC Supplier No. HYW01362710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trans,trans-2,4-Decadienal, also known by its synonym (2E,4E)-癸-2,4-二烯醛, is a lipid peroxidation product derived from linoleic acid. This compound has a molecular weight of 152.23 and the chemical formula C10H16O. It is a liquid with a density of 0.854±0.06 g/cm3, appearing light yellow to green yellow. Primarily used for scientific research or pharmaceutical registration.

  • Derived from linoleic acid
  • Molecular weight of 152.23
  • Light yellow to green yellow liquid
  • Used for scientific research or pharmaceutical registration
  • Metabolized via oxidation or glutathione conjugation
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at -20°C for pure form; -80°C or -20°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.