Learn More
Medchemexpress LLC trans,trans-2,4-Decadienal | 25152-84-5 | 99.9% | 152.23 | 5 G
SDP
Catalog No. 5000454219
Encompass_Preferred
trans,trans-2,4-Decadienal is a lipid peroxidation product of linoleic acid. Two metabolic pathways for its biotransformation in vivo are proposed: oxidation to the corresponding carboxylic acid in liver cells, and glutathione conjugation, breakdown, and aldehyde reduction, generating cysteine-conjugated 2,4-decadien-1-ol in both liver and lung cells.
  • Oxidation to 2,4-decadienoic acid in liver cells.
  • Glutathione (GHS) conjugation, GSH breakdown, and aldehyde reduction.
  • Generates cysteine-conjugated 2,4-decadien-1-ol in both liver and lung cells.
Catalog No. 50-004-54219 Supplier Medchemexpress LLC Supplier No. HYW0136275G
Add to Cart
Add to Cart
trans,trans-2,4-Decadienal is a lipid peroxidation product of linoleic acid. Two metabolic pathways for its biotransformation in vivo are proposed: oxidation to the corresponding carboxylic acid in liver cells, and glutathione conjugation, breakdown, and aldehyde reduction, generating cysteine-conjugated 2,4-decadien-1-ol in both liver and lung cells.
  • Oxidation to 2,4-decadienoic acid in liver cells.
  • Glutathione (GHS) conjugation, GSH breakdown, and aldehyde reduction.
  • Generates cysteine-conjugated 2,4-decadien-1-ol in both liver and lung cells.