Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-2,4-Heptadienal 90.0+%, TCI America™
SDP

Catalog No. H06815ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06815ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06815ML Supplier TCI America Supplier No. H06815ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4313-03-5
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00007005
InChI Key SATICYYAWWYRAM-VNKDHWASSA-N
PubChem CID 5283321
IUPAC Name (2E,4E)-hepta-2,4-dienal
SMILES CCC=CC=CC=O

Specifications

Boiling Point 85°C
Quantity 5 mL
UN Number 2810
Formula Weight 110.16
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material trans,trans-2,4-Heptadienal
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.