Learn More
Medchemexpress LLC Trans,trans-2,4-hexadien-1-ol | 17102-64-6 | MFCD00002925 | >=97.0% | 98.14 g/mol | C6H10O | 5 G
SDP
Catalog No. 5000299717
Encompass_Preferred
(2E,4E)-Hexa-2,4-dien-1-ol (trans,trans-2,4-hexadien-1-ol) is a small unsaturated aliphatic alcohol used as a research reagent and synthetic intermediate. It is commonly supplied stabilized to reduce oxidation and is employed in organic synthesis and analytical applications. Key identifiers include CAS 17102-64-6 and formula C6H10O.
  • Unsaturated aliphatic alcohol for organic synthesis and research
  • Typically supplied stabilized to limit oxidation
  • Typical purity ≥97.0%
  • Chemical formula C6H10O; molecular weight 98.14 g/mol
  • Suitable for small-scale laboratory and analytical use
Catalog No. 50-002-99717 Supplier Medchemexpress LLC Supplier No. HYW0176905G
(2E,4E)-Hexa-2,4-dien-1-ol (trans,trans-2,4-hexadien-1-ol) is a small unsaturated aliphatic alcohol used as a research reagent and synthetic intermediate. It is commonly supplied stabilized to reduce oxidation and is employed in organic synthesis and analytical applications. Key identifiers include CAS 17102-64-6 and formula C6H10O.
  • Unsaturated aliphatic alcohol for organic synthesis and research
  • Typically supplied stabilized to limit oxidation
  • Typical purity ≥97.0%
  • Chemical formula C6H10O; molecular weight 98.14 g/mol
  • Suitable for small-scale laboratory and analytical use