Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans,trans-2,4-Hexadien-1-ol

Catalog No. 18305910G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-305-910G 10 g
18-305-950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-305-910G Supplier Sigma Aldrich Supplier No. 18305910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans,trans-2,4-Hexadien-1-ol

Specifications

CAS 17102-64-6
Melting Point 28°C to 33°C (lit.)
Density 0.871 g/mL (at 25°C (literature))
Boiling Point 80°C (12 mmHg)
Molecular Formula C6H10O
Refractive Index n20/D 1.5 (literature)
Linear Formula CH3CH=CHCH=CHCH2OH
MDL Number MFCD00002925
Quantity 10 g
Synonym Sorbic alcohol
Molecular Weight (g/mol) 98.14
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.