Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans,trans-2,4-Hexadienoic acid potassium salt

Catalog No. 3597691KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-976-91KG 1 kg
35-976-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-976-91KG Supplier Sigma Aldrich Supplier No. 3597691KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sorbates have been reported to be less toxic than benzoate and have been classified as generally recognized as safe (gras) additives by the u.s
  • Food and drug administration (fda).sorbic acid is metabolised to mainly to carbon dioxide
  • While the minor amounts are converted to trans,trans-muconic acid (ttma), which is excreted unchanged into the urine
  • Urinary ttma is a biomarker for the occupational and environmental exposure to benzene
  • Ability of trans,trans-2,4-hexadienoic acid potassium salt (sorbic acid potassium salt, potassium sorbate) to induce chromosome aberrations, sister chromatid exchanges (sce) and gene mutations in cultured chinese hamster v79 cells has been examined.potassium sorbate is reported to be less genotoxic than the sodium salt analog.reagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 24634-61-5
Molecular Formula C6H7KO2
Linear Formula CH3CH=CHCH=CHCOOK
MDL Number MFCD00016546
Quantity 1 kg
Synonym Potassium sorbate; Potassium 2,4-hexadienoate; Sorbic acid potassium salt
Molecular Weight (g/mol) 150.22
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.