Learn More
Medchemexpress LLC trans trans-2 4-Nona 100g
SDP
Supplier:  Medchemexpress LLC HYW127521100G
Encompass_Preferred
Trans trans-2 4-NonadienalIt is an organic compound belonging to aldehydes It has a strong pungent smell and is commonly found in a variety of foods including fruits and vegetables Trans trans-2 4-NonadienalIt has a variety of applications in the flavor and fragrance industry especially as a fragrance agent in products such as perfume cologne and air fresheners In addition it can also be used as an intermediate in the synthesis of various chemicals and drugs
Catalog No. 50-002-04901