Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™
SDP

Catalog No. D47971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47971G 1 g
D47975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47971G Supplier TCI America Supplier No. D47971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82832-57-3
Molecular Formula C21H30F2
Molecular Weight (g/mol) 320.468
MDL Number MFCD09838997
InChI Key FSWZOZXLWVWJAH-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl
PubChem CID 581325
IUPAC Name 1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
SMILES CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 44°C
Color White
Quantity 1 g
Formula Weight 320.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4′-propylbicyclohexyl
EINECSNumber (4)-1883
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.