Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4'-Butyl-4-(3,4-difluorophenyl)bicyclohexyl 98.0+%, TCI America™
SDP

Catalog No. B49255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49255G 5 g
B492525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49255G Supplier TCI America Supplier No. B49255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82832-58-4
Molecular Formula C22H32F2
Molecular Weight (g/mol) 334.495
MDL Number MFCD13188634
InChI Key NZXZINXFUSKTPH-UHFFFAOYSA-N
Synonym trans,trans-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4-butylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-butyl-bicyclohexyl, 4-4-4-butylcyclohexyl cyclohexyl-1,2-difluorobenzene, 4-trans,trans-4'-butyl 1,1'-bicyclohexyl-4-yl-1,2-difluorobenzene, 4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, 1s,1'r,4r,4'r-4-butyl-4'-3,4-difluorophenyl-1,1'-bi cyclohexane, trans,trans-4'-butyl-4-3,4-difluorophenyl bicyclohexyl, trans,trans-4-butyl-4-3,4-difluorophenyl bicyclohexyl
PubChem CID 14993439
IUPAC Name 4-[4-(4-butylcyclohexyl)cyclohexyl]-1,2-difluorobenzene
SMILES CCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 47°C
Color White
Quantity 5 g
Formula Weight 334.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-Butyl-4-(3,4-difluorophenyl)bicyclohexyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.