Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4'-Ethyl-4-(4-trifluoromethoxyphenyl)bicyclohexyl 98.0+%, TCI America™
SDP

Catalog No. E11655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11655G Supplier TCI America Supplier No. E11655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135734-59-7
Molecular Formula C21H29F3O
Molecular Weight (g/mol) 354.457
MDL Number MFCD09842837
InChI Key PBIFTROFCZATFC-UHFFFAOYSA-N
PubChem CID 604782
IUPAC Name 1-[4-(4-ethylcyclohexyl)cyclohexyl]-4-(trifluoromethoxy)benzene
SMILES CCC1CCC(CC1)C2CCC(CC2)C3=CC=C(C=C3)OC(F)(F)F

Specifications

Melting Point 73°C
Color White
Quantity 5 g
Formula Weight 354.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-Ethyl-4-(4-trifluoromethoxyphenyl)bicyclohexyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.