Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4'-Ethylbicyclohexyl-4-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. E12515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12515G Supplier TCI America Supplier No. E12515G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84976-67-0
Molecular Formula C15H26O2
Molecular Weight (g/mol) 238.371
MDL Number MFCD05864406
InChI Key OQIHEFMTIUJJET-UHFFFAOYSA-N
Synonym trans,trans-4′C-Ethyl-1,1′C-bicyclohexane-4-carboxylic Acid, trans-4-(trans-4-Ethylcyclohexyl)cyclohexanecarboxylic Acid
PubChem CID 11694375
IUPAC Name 4-(4-ethylcyclohexyl)cyclohexane-1-carboxylic acid
SMILES CCC1CCC(CC1)C2CCC(CC2)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 238.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-Ethylbicyclohexyl-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.