Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans,trans-dibenzylideneacetone | 35225-79-7 | 99.9% | 234.30 | 5 G
SDP

Catalog No. 5000452763
Encompass_Preferred

trans,trans-Dibenzylideneacetone, also known as (E,E)-Dibenzylideneacetone or trans,trans-Dibenzalacetone, is a dibenzylideneacetone compound. It exhibits weak antimalarial activity, shown to inhibit the *P. falciparum* 3D7 strain with an IC50 of 213.41 μM.

  • High purity of 99.87%
  • Solid, light yellow to yellow appearance
  • Molecular weight of 234.30
  • Chemical formula C17H14O
  • Stable for 3 years at room temperature
  • Inhibits *P. falciparum* 3D7 strain

Catalog No. 50-004-52763 Supplier Medchemexpress LLC Supplier No. HYW0121925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans,trans-Dibenzylideneacetone, also known as (E,E)-Dibenzylideneacetone or trans,trans-Dibenzalacetone, is a dibenzylideneacetone compound. It exhibits weak antimalarial activity, shown to inhibit the *P. falciparum* 3D7 strain with an IC50 of 213.41 μM.

  • High purity of 99.87%
  • Solid, light yellow to yellow appearance
  • Molecular weight of 234.30
  • Chemical formula C17H14O
  • Stable for 3 years at room temperature
  • Inhibits *P. falciparum* 3D7 strain

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.