Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans,trans-Farnesol

Catalog No. 2775411G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-754-11G 1 g
27-754-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-754-11G Supplier Sigma Aldrich Supplier No. 2775411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans,trans-farnesol is sesquiterpene alcohol, which is commonly found in many essential oils, citrus fruits, propolis, and plant-derived foods and beverages. it is an effective bioactive agent that can be used in pharmaceuticals and cosmetics.trans,trans-farnesol has been used in the synthesis of:
  • Cecropia juvenile hormone
  • Terpenes, such as squalenes, cembranoids and forskolintandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. 277541.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin

Specifications

CAS 106-28-5
Density 0.879 g/mL (at 25°C (literature))
Boiling Point 137°C (3 mmHg)
Molecular Formula C15H26O
Linear Formula (CH3)2 C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)=CHCH2OH
Refractive Index n20/D 1.490 (literature)
MDL Number MFCD00002918
Quantity 1 g
Synonym (E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol; trans,trans-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol
Molecular Weight (g/mol) 222.37
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.