Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-Farnesyl pyrophosphate ammonium salt, Analytical Standard, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111003600 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3600 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3600 Supplier MilliporeSigma Supelco Supplier No. 447225MG
Add to Cart
Add to Cart

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
Physical Form Neat
Linear Formula C15H28O7P2 · xNH3
Synonym (E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl pyrophosphate ammonium salt; Farnesyl diphosphate ammonium salt
Quantity 5 mg
Formula Weight 382.33 (free acid basis)
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.