Learn More
Medchemexpress LLC trans-Urocanic acid | 3465-72-3 | 99.9% | 138.12 | 5 G
SDP
Catalog No. 5000439705
Encompass_Preferred
trans-Urocanic acid (trans-UCA) is a natural epidermal constituent known for regulating immune function. It inhibits human natural killer cell (NK) activity in vitro and partially inhibits the cytotoxic function of IL-2-activated NK cells. It also reduces IL-2-induced activation of NK cells.
  • Natural epidermal constituent
  • Inhibits human natural killer cell (NK) activity in vitro
  • Regulates immune function
  • Partially inhibits cytotoxic function of IL-2-activated NK cells
  • Reduces IL-2-induced activation of NK cells
Catalog No. 50-004-39705 Supplier Medchemexpress LLC Supplier No. HY1130085G
trans-Urocanic acid (trans-UCA) is a natural epidermal constituent known for regulating immune function. It inhibits human natural killer cell (NK) activity in vitro and partially inhibits the cytotoxic function of IL-2-activated NK cells. It also reduces IL-2-induced activation of NK cells.
  • Natural epidermal constituent
  • Inhibits human natural killer cell (NK) activity in vitro
  • Regulates immune function
  • Partially inhibits cytotoxic function of IL-2-activated NK cells
  • Reduces IL-2-induced activation of NK cells