Learn More
Medchemexpress LLC HY-113427 25mg Medchemexpress, trans-Vaccenic acid CAS:693-72-1 Purity:>98%
SDP
Catalog No. 501932463
Encompass_Preferred
Medchemexpress, HY-113427 25mg trans-Vaccenic acid CAS:693-72-1 trans-Vaccenic acid is a precursor for the synthesis of saturated fatty acid in the rumen and of conjugated linoleic acid (CLA) at the tissue level. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2463 Supplier Medchemexpress LLC Supplier No. HY11342725MG
Medchemexpress, HY-113427 25mg trans-Vaccenic acid CAS:693-72-1 trans-Vaccenic acid is a precursor for the synthesis of saturated fatty acid in the rumen and of conjugated linoleic acid (CLA) at the tissue level. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.