Learn More
Gold Biotechnology Inc trans Zeatin 1 g
SDP
Catalog No. 5000562071
Encompass_Preferred
trans -Zeatin is a naturally occurring cytokinin plant growth regulator routinely used in plant tissue culture to induce cell division and shoot formation trans -zeatin is freely soluble in acetic acid and water
Catalog No. 50-005-62071 Supplier Gold Biotechnology Inc Supplier No. Z1051
trans -Zeatin is a naturally occurring cytokinin plant growth regulator routinely used in plant tissue culture to induce cell division and shoot formation trans -zeatin is freely soluble in acetic acid and water