Learn More
Medchemexpress LLC trans-Zeatin | 1637-39-4 | 99.6% | 219.24 | 1 ML
SDP
Catalog No. 5000401693
Encompass_Preferred
trans-Zeatin is a plant cytokinin that plays a vital role in cell growth, differentiation, and division. It also acts as an inhibitor of UV-induced MEK/ERK activation.
  • Plant cytokinin
  • Inhibits UV-induced MEK/ERK activation
  • Plays a role in cell growth, differentiation, and division
  • Available in solid and solution forms
Catalog No. 50-004-01693 Supplier Medchemexpress LLC Supplier No. HY1970010MM/1ML
trans-Zeatin is a plant cytokinin that plays a vital role in cell growth, differentiation, and division. It also acts as an inhibitor of UV-induced MEK/ERK activation.
  • Plant cytokinin
  • Inhibits UV-induced MEK/ERK activation
  • Plays a role in cell growth, differentiation, and division
  • Available in solid and solution forms