Learn More
Medchemexpress LLC trans-Zeatin | 1637-39-4 | 99.6% | 219.24 | 5 MG
SDP
Catalog No. 5000401331
Encompass_Preferred
trans-Zeatin is a plant cytokinin that is important in cell growth, differentiation, and division. It also inhibits UV-induced MEK/ERK activation.
  • Plant cytokinin
  • Plays an important role in cell growth, differentiation, and division
  • Inhibits UV-induced MEK/ERK activation
Catalog No. 50-004-01331 Supplier Medchemexpress LLC Supplier No. HY197005MG
trans-Zeatin is a plant cytokinin that is important in cell growth, differentiation, and division. It also inhibits UV-induced MEK/ERK activation.
  • Plant cytokinin
  • Plays an important role in cell growth, differentiation, and division
  • Inhibits UV-induced MEK/ERK activation