Learn More
Selleck Chemical LLC Trans-Zeatin S4884-5mg
SDP
Catalog No. 503109210
Encompass_Preferred
Trans-Zeatina ((E)-Zeatin) is the member of the plant growth hormone family known as cytokinins which regulate cell division development and nutrient processing Trans-Zeatin inhibits UVB-induced MMP-1 expression c-Jun activation and phosphorylation of ERK JNK and p38 MAP kinases (MAPKs) dose-dependently
Catalog No. 50-310-9210 Supplier Selleck Chemical LLC Supplier No. S48845MG
Trans-Zeatina ((E)-Zeatin) is the member of the plant growth hormone family known as cytokinins which regulate cell division development and nutrient processing Trans-Zeatin inhibits UVB-induced MMP-1 expression c-Jun activation and phosphorylation of ERK JNK and p38 MAP kinases (MAPKs) dose-dependently