Learn More
Medchemexpress LLC trans-Zeatin-d5 | 72963-19-0 | 99.2% | 224.27 | 1 MG
SDP
Catalog No. 5000400915
Encompass_Preferred
trans-Zeatin-d5 is deuterium labeled trans-Zeatin, a plant cytokinin vital for cell growth, differentiation, and division. It also inhibits UV-induced MEK/ERK activation. This compound can serve as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules primarily assists in quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled trans-Zeatin
  • Plays an important role in cell growth, differentiation, and division
  • Inhibits UV-induced MEK/ERK activation
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-00915 Supplier Medchemexpress LLC Supplier No. HY19700S1MG
trans-Zeatin-d5 is deuterium labeled trans-Zeatin, a plant cytokinin vital for cell growth, differentiation, and division. It also inhibits UV-induced MEK/ERK activation. This compound can serve as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules primarily assists in quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled trans-Zeatin
  • Plays an important role in cell growth, differentiation, and division
  • Inhibits UV-induced MEK/ERK activation
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs